Learn More
Bulbtronics™ General Electric™ Halogen Lamp: MR16
Manufacturer:  Bulbtronics™ 001404
 View more versions of this product
Catalog No. 14-395-492
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
GX5.3/2 Pin
3250K
5.08cm
Microscope, surgical lights, Fiber optic light source
4.44cm
MR16 Shape
150w
Halogen Lamp, Faceted Dichroic Reflector
Base down to horizontal
2 in.
CC6/Coiled Coil Transverse
1.75 in.
250 hr.
21V
Halogen