Learn More
Bulbtronics™ Germicidal Lamp
Manufacturer:  Bulbtronics, Inc. 000179
 View more versions of this product
Catalog No. 14-395-341
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
G13/Medium Bi Pin
3.17cm
119.8cm
5000 hr.
40w
Linear Double Ended Lamp
1.25 in.
47.1 in.
Germicidal
T10 Shape
254nm