Bulbtronics Halogen Light Bulbs

Manufacturer: Bulbtronics, Inc. 0000938

 View more versions of this product

Catalog No. 14-395-871

  Check Availability
Add to Cart

Alert

The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.


    Specifications

    Color Clear
    Diameter (English) Outer 2 in.
    Lighting Life 50 hr.
    Voltage 24V
    Wattage 250w
    Light Source Halogen
    Product Type Halogen Bulb
    Base Style GX5.3/2 Pin
    Diameter (Metric) Outer 5.08cm
    Filament C8/Single Coil Axial
    For Use With (Equipment) Fiber optic light source
    Length (English) 1.75 in.
    Length (Metric) 4.4cm
    Shape MR16 Shape

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit