Bulbtronics Incandescent Light Bulbs

Manufacturer: Bulbtronics, Inc. 002747

 View more versions of this product

Catalog No. 14-395-598

  Check Availability
Add to Cart

Alert

The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.


    Specifications

    Base Style P30D/DC Prefocus
    Color Clear
    Diameter (English) Outer 1 in.
    Diameter (Metric) Outer 2.54cm
    Filament C6/Single Coil Transverse
    Length (English) 2 in.
    Length (Metric) 5.1cm
    Lighting Life 100 hr.
    Shape S8 Shape
    Voltage 6.5V
    Wattage 17.8w
    Light Source Incandescent
    Product Type Incandescent Lamp

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit