Learn More
Bulbtronics™ Incandescent Light Bulbs
Manufacturer:  Bulbtronics™ 002753/DEL
 View more versions of this product
Catalog No. 14-395-600
Add to cart
Alert:
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Specifications
Specifications
E12/Candelabra
C7A
5.71cm
S11 Shape
25w
Incandescent Lamp
Clear
2.25 in.
500 hr.
130V
Incandescent