Learn More
Bulbtronics™ Lumen Dynamics Group Mercury Arc Bulb
For fluorescence microscopy
Manufacturer:  Bulbtronics™ 0064381/DEL
 View more versions of this product
Catalog No. 14-395-231
Add to cart
Description
Description
Used in all X-Cite 120-model illuminators
  • 120w AC or 200w DC
  • Base: Wire leads with connectors
  • Bulb Life: 2000 hr.
Specifications
Specifications
Wire leads with connectors
2000 hr.
Mercury Arc
X-Cite exacte illuminator
200w
Mercury Arc Bulb