Learn More
Bulbtronics™ Narva™ Halogen Bulbs
Manufacturer:  Bulbtronics™ 000386/DEL
 View more versions of this product
Catalog No. 14-395-383
Add to cart
Specifications
Specifications
PY16-1.25
0.37 in.
Microscope
1.18 in.
2.99cm
510lm
6V
Halogen
Clear
0.95cm
0.049 in.
0.12cm
100 hr.
T3 Shape
25w
Halogen Lamp