Learn More
Bulbtronics™ Ushio Halogen Lamp: T3
Manufacturer:  Bulbtronics™ 001740/DEL
 View more versions of this product
Catalog No. 14-395-525
Add to cart
Specifications
Specifications
GY6.35/2 Pin
3300K
0.43 in.
CBAR6/C6F/Single Coil Flat/Oval Transverse
1.18 in.
2.9cm
50 hr.
T3.5 Shape
100w
Halogen Lamp
Base down to horizontal
Clear
1.09cm
Microscope, collimator
1.73 in.
4.39cm
3400lm
12V
Halogen