Learn More
Bullard™ 41 Series Breathing AirLine Filter Accessory, Pressure Relief Valve
SDP
Replacement part for use with 41 Series Breathing AirLine Filters
Supplier:  Bullard™ 41PRV
Catalog No. 19-824-405
Add to Cart
Alert:
The 41 Series Airline filters do not remove carbon monoxide and other toxic gases.
Description
Description
  • For 41P2, 41P2E, and 41P6/41P6WM airline filters
  • Set at 125 psig (8.6 bar)
Specifications
Specifications
41 Series Breathing AirLine Filter
41PRV
125 psi
Pressure relief valve