Learn More
Bullard™ 41 Series Breathing AirLine Filter Accessory, Regulator Gauge
Replacement part for use with 41 Series Breathing AirLine Filters
Supplier: Bullard™ 41RG
Alert:
The 41 Series Airline filters do not remove carbon monoxide and other toxic gases.
Description
- For 41P2, 41P2E, and 41P6 airline filters
Specifications
41RG | |
Pressure Gauge |