Learn More
Bullard™ Inlet and Outlet Filters for Electric Free-Air™ Pumps
SDP
For use with Free-Air pumps
Supplier:  Bullard™ S17101
 View more versions of this product
Catalog No. 19-021-308
Add to Cart
Description
Description
Coupler Splitter/Adapter: Converts coupler to double 1/2 in. Industrial Interchange female couplers (one flow through, one shut-off)
Inlet Filter: Change every 500 hours or as required
Outlet Filter: Carbofine; change every 200 hours or as required
Specifications
Specifications
S17101
Outlet filter
Product Suggestions
Product Suggestions