Bullard™ Classic Series C30 Hardhats
Offer a comfortable and secure fit
Manufacturer: Bullard Company 30FGP
 View all options for this product
Catalog No. 19-021352
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Certifications/Compliance Meets ANSI Z89.1-2003; Type I; Class G and E.
    Color Green
    Material HDPE,HDPE
    Suspension Type 6-Point Standard,6-Point Standard
    • Wide profile and brim increase protection from sun and improve ventilation
    • Constructed of high-density polyethylene
    • Accessory slots accept most attachments, including welding helmets, earmuffs, faceshields, and communication devices
    • Six-point suspension is either a standard design with sizing posts or a Flex-Gear™ ratchet design with sizing knob
    • Generous cotton browpad is absorbent and fast drying; brow pad is replaceable
  • Description & Specifications
  • Accessories
Related Products
Product Content Feedback