Learn More
Bullard™ 41 Series CO Monitor and Filter
Provides 7-stage filtration combined with carbon monoxide (CO) monitoring.
Supplier: Bullard™ COHP41P6
Description
Monitors and filters may also be used to supply drier, cleaner air to tools and pneumatic systems to reduce downtime and repair costs
7-Stage Filtration
- Water is removed by condensation of air in outer cylinder
- Carded cotton removes particulates
- Activated alumina absorbs oil and moisture
- Activated charcoal removes odors and moisture
- Felt material removes particulates
- Carded cotton removes particulates
- Respiratory felt at final stage acts as a final filter before air is transferred to worker or air driven tools
- Steel construction for rugged industrial environments
- Large disposable filter cartridge for long life
- Carrying handle available for easy transport
- Weighs only 23 pounds
- Two or six outlets
- Free standing or wall mountable
Warranty and Services
One-year limited warranty
Specifications
6 outlet | |
41 Series Airline Respirator | |
7-stage filtration filter unit, CO monitor with auto calibration, audible and visual alarms for high co and low battery and 6 ft. 110 V power cord, pressure gauge, 125 psig relief valve and pressure regulator | |
23.5 in. | |
22 lb. | |
CO Monitor and Filter Kit |
75 cfm | |
Steel | |
COHP41P6 | |
60 cm | |
10 kg |