Learn More
Bullard™ Evolutionary Air Hazardous Location (EVAHL) PAPR System Accessory, Double Bib Hood
Designed to be used with the EVAHL Powered Air Purifying Respirator
Supplier: Bullard™ EVAHLRT3T
View more versions of this product
Safety Services Available
Overview
PPE Assessments
Description
- Ideal for painting, chemical handling, healthcare, pharmaceutical applications
- Choice of hood configurations
- Airflow rate: 8.5/7 cfm
- 77dBA low voltage and 77dBA low flow alarm
- Lithium ion polymer battery (1-year life span)
Certifications
ISO 9001, NIOSH, CSA, Class I Division 2, Class II Division 2, Class III
Specifications
Lithium Ion Polymer | |
ISO 9001, NIOSH, CSA, Class I Division 2, Class II Division 2, Class III | |
Double Bib Suspension-free Hood | |
Suspension-free |