Learn More
Bullard™ Evolutionary Air Hazardous Location (EVAHL) PAPR System Accessory, QC Hood
Designed to be used with the Evolutionary Air Hazardous Location Powered Air Purifying Respirator
Supplier: Bullard™ EVAHL20TICTDB
View more versions of this product
Safety Services Available
Overview
PPE Assessments
Description
- Dupont™ Tychem QC hood
- Double bib with ratchet suspension
- Sealed seams (Cat. No. 17989516 only)
- Threaded connector style
Certifications
ISO 9001, NIOSH, CSA, Class I Division 2, Class II Division 2, Class III
Specifications
Lithium Ion Polymer | |
Double Bib Hood | |
Vinyl decon belt, lamp connector style/threaded connector style |
ISO 9001, NIOSH, CSA, Class I Division 2, Class II Division 2, Class III | |
Ideal for painting, chemical handling, healthcare, pharmaceutical | |
Ratchet suspension |