Bullard™ EVAHL PAPR System Accessory, QC Hood
Designed to be used with the Evolutionary Air Hazardous Location Powered Air Purifying Respirator
Manufacturer: Bullard Company EVAHL20TPT
 View more versions of this product
Catalog No. 17-989-524
  Check Availability
Add to Cart
    Specifications
    Product Type Single bib hood
    For Use With (Application) Ideal for painting, chemical handling, healthcare, pharmaceutical
    Battery Type Lithium Ion Polymer
    Suspension Type Ratchet suspension
    Includes Solvent Resistant Lens, clamp connector style/threaded connector style
    Certifications ISO 9001, NIOSH, CSA, Class I Division 2, Class II Division 2, Class III
    Battery Life 1 Year
  • Description & Specifications
Related Products
Product Content Feedback