Learn More
Bullard™ Firedome™ FXA-1 Helmet
Engineered to meet U.S. Navy requirements for shipboard firefighter helmets
Supplier: Bullard™ FXA1RED
Description
Features ear/neck protector made of three separate materials and special impact liner.
- Tough outer shell of resin-bonded fiberglass provides more resistance to chemical attack and more stability at extreme temperatures than thermoplastic models
- 3M™ Scotchlite™ striping resists edge curl and retains reflectivity to 500°F, making it the toughest material available for helmet markings
- Sure-Lock™ ratchet headband adjusts to wearer's head with a quick turn of the knob
- Vertical adjustment hanger keys on front and rear positions of headband provide a truly customized fit
- Browpad is washable and replaceable
- 4 in. PPC, optically correct faceshield pivots a full 90° above the outer shell for optimum interface with SCBA facepieces
Order Info
Available through Fisher Safety's GSA Advantage!™ Schedule 84. Gold-coated faceshield, aluminized helmet cover and aluminized shroud available; contact your Fisher Safety Customer Service Representative for ordering information.
Compliance
Meets NFPA 1971 Standards on Protective Ensemble for Structural Fire Fighting, 2013 Edition.
Specifications
Meets NFPA 1971 Standards on Protective Ensemble for Structural Fire Fighting, 2007 Edition. | |
Red | |
Urethane |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.