Learn More
Bullard™ Constant Flow Assemblies for RT Series Airline Respirators
Flow control assembly for use with RT Series respirator, does not include breathing tube.
Supplier: Bullard™ F30
View more versions of this product
Safety Services Available
Overview
PPE Assessments
Description
- Constant flow accessory
- For use with RT series airline respirator
- Flow control assemblies
- Nipple types include bayonet, CEJN, industrial interchange, schrader, or snap-tite
- Does not include breathing tube
Specifications
0.25 in. Industrial Interchange (Hansen-compatible) Nipple | |
Flow Control Assembly | |
CC20 Series Respirators | |
F30 |