Bullard™ PA30 Series Complete PAPR Assemblies
Choose from decon-friendly hospital preparedness design, or any one of numerous intrinsically safe models
Manufacturer: Bullard Company PA30ISLFLDB
 View more versions of this product
Catalog No. 19-168-310
  Check Availability
Add to Cart
    Specifications
    Certifications NIOSH
    For Use With (Equipment) Pa30 powered air purifying respirator system
    For Use With (Application) HEPA, Organic Vapor, Acid Gas Protection
    Includes One battery, Decon Belt, PA3 blower, breathing tube, one loose-fitting facepiece, quick charger, low voltage alarm and air flow indicator
    Airflow Rate >7cfm
    Battery Life 6-7 hour run time
    Product Type 20LFL Loose-Fitting Facepiece System with Decon Belt
  • Description & Specifications
Related Products
Product Content Feedback