Learn More
Bullard PA30 Series Complete PAPR Assemblies
Choose from decon-friendly hospital preparedness design, or any one of numerous intrinsically safe models
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
19-167-755
|
Bullard
PA30LFLSB |
20LFL Loose-Fitting Facepiece System with Standard Comfort Belt |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
Description
Sold as complete assemblies. For descriptions of individual components, see separate entries.
Hospital Preparedness System
- Comes with Tychem SL hood, protective breathing tube cover, vinyl decon belt, and one set of NBC canisters Smooth, “decon-friendly” housing is sealed and gasketed, with downward slopes for easy runoff
- Rechargeable NiMH battery packs are engineered inside the profile of the housing to provide even weight distribution and eliminate battery cables that may entangle or short One battery lasts up to five hours 77dBA audible low-voltage alarm inside breathing tube connector sends signal to the headphones Airflow: >8cfm
- All models feature PA3IS intrinsically safe blower with battery, breathing tube, quick charger, low-voltage alarm, and airflow indicator Unit with solvent-resistant lens suits painting applications
- RT1 and RT2 hoods with extra-large lens provide high-performance results using an inflatable neck cuff design that provides positive custom seal around user's neck For more information about individual hood features and benefits, see separate descriptions Battery provides 6-7 hours of run time
- Airflow: >7cfm