Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bullard™ Replacement Filter for Calibration Kits for 15 & 30 CFM Models

For calibration kits for 15 and 30 CFM models.

Manufacturer:  Bullard™ CAB30FEA

 View more versions of this product

Catalog No. 19-100-680


  Check Availability
Add to cart

Specifications

Description

For calibration kits for 15 and 30 CFM models.
Description & Specifications

Specifications

Element A-Particulate
Removes 95% of solid contaminants and bulk liquids, down to 5μ from the breathing air system
Replacement Filter
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit