Bullard™ Replacement Filter for Calibration Kits for 50 CFM Models
For calibration kits for 50 CFM models.
Manufacturer: Bullard™ CAB50FK
 View more versions of this product
Catalog No. 19-100-687
  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    Includes Element A-Particulate, Element B-Coalescing, Element C-Activated Charcoal
    For Use With (Application) Removes 95% of solid contaminants, bulk liquids down to 5μ, particulates, extremely fine oil mists down to 0.01μ with 99.9% filter efficiency, unpleasant oil and hydrocarbon vapors and odors from the breathing air system
    Product Type Replacement Filter
    For calibration kits for 50 CFM models.
  • Description & Specifications
Related Products
Product Content Feedback