Learn More
Bullard™ Replacement Parts for PA30IS Series Powered Air Purifying Respirators: HEPA Filter
Replacement Parts for use with PA30IS Series Powered Air Purifying Respirators
Supplier: Bullard™ PAPRFC1
Safety Services Available
Overview
PPE Assessments
Description
Replacement Parts for use with PA30IS Series Powered Air Purifying Respirators
Specifications
NIOSH | |
Hepa Filter | |
Pa30is series powered air purifying respirator |
Magenta | |
99.97% for 0.3μ | |
6/Pk. |