Bullard™ Standard Series S62 Vented Hardhats
Low-profile shell design with four-point suspension system
Manufacturer: Bullard Company 62WHP
 View all options for this product
Catalog No. 19-160-457
  Check Availability
Add to Cart
    Certifications/Compliance Meets ANSI Z89.1-2003; Type I; Class E and G (S61 Model); Class C (S62 Vented).
    Color White
    Suspension Type 4-Point Pin-Lock,4-Point Pin-Lock
    Setting the Standard. Sleek, low-profile design with a patented Flex-Gear ratchet suspension option.
    • Absorbent, replaceable cotton brow pad is pillowed for maximum comfort
    • Ideal for construction and everyday work environments
    • One of the lightest helmets of its class
    • Vented, providing increased ventilation and comfort
    • Accessory slots allow attachment of accessories such as earmuffs and face protection
    • Vertical height adjustment capability (2 front, 2 rear)
    Pinlock or Ratchet Suspension
    • Choose 4-point pinlock suspension or 4-point Flex-Gear ratchet suspension
    • Pinlock suspension provides incremental sizing of 1/16 hat size (5mm), for a secure fit
    • Ratchet suspension can be adjusted between hat sizes 6 1/2-8
  • Description & Specifications
  • Accessories
Related Products
Product Content Feedback