Bullard™ Supplied-Air Respirator Accessories: Breathing Tube Clamps

For Supplied-Air Respirator

Manufacturer: Bullard Company S18051BLACK

 View more versions of this product

Catalog No. 19-824-426

  Check Availability
Add to Cart

    Specifications

    For Use With (Equipment) Breathing tubes of pa20 series powered air purifying respirator, supplied air respirator
    Product Type Hose Clamp
    Includes 10 Clamps
    Material Nylon
    Color Black

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit