Bullard™ Tychem™ QC Hoods for Respirators

For use with CC20 Airline Respirators and EVA, EVAHL, PA20, PA30, PA30IS PAPRs

Manufacturer: Bullard Company 20TPN

 View more versions of this product

Catalog No. 18-998-035

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Quantity 10 Pack
    For Use With (Equipment) Cc20™ series airline respirator and eva™, evahl, pa20, pa30, pa30is powered air purifying respirator,Cc20™ series airline respirator and eva™, evahl, pa20, pa30, pa30is powered air purifying respirator
    Product Type Hood
    For Use With (Application) Painting
    Includes 9 in. l bib, solvent resistant clear petg lens, headband
    Material Tychem™ QC
    Certifications NIOSH
    Color White

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit