Learn More
Bullard™ Tychem™ SL Hoods for Respirators — Vinyl Lens, 19in. (48.3cm) Inner Bib
For use with CC20 Airline Respirators and PA20, PA30, PA30IS, EVA, EVAHL PAPRs
Manufacturer:  Bullard™ 20SICVN
 View more versions of this product
Catalog No. 19-065-767
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Tychem SL is Tychem coated with Saranex™ 23-P to provide protection from an even wider range of chemicals. Hoods cover head, neck, and face; they feature a wide-angled plastic lens.
Double Bib (Inner Bib) Style: Outer bib with a long inner bib; tuck inner bib into shirt or coveralls to funnel excess air from the hood to the body
Double Bib (Inner Bib) Style for Use with Hardhat: Accommodates Bullard hardhats
Specifications
Specifications
PA20 and PA30 PAPR and CC20 Airline Respirators
Safety
Vinyl lens
Tychem™ SL Coated with Saranex™ 23-P, Vinyl Lens
Double Bib Hood
No suspension