Bullard™ Tychem™ SL Hoods for Respirators — Vinyl Lens, 19 in. (48.3cm) Inner Bib
For use with CC20 Airline Respirators and PA20, PA30, PA30IS, EVA, EVAHL PAPRs
Manufacturer: Bullard Company 20SICVH
 View more versions of this product
Catalog No. 19-065-768
  Check Availability
Add to Cart
    Specifications
    For Use With (Equipment) PA20, PA30, PA30IS, EVA, EVAHL PAPRs and CC20 Airline,PA20, PA30, PA30IS, EVA, EVAHL PAPRs and CC20 Airline
    Product Type Double Bib Hood for hard hats
    Material Tychem™ SL Coated with Saranex™ 23-P
    Size Extra Long
    Material (Lens) Vinyl
  • Description & Specifications
Related Products
Product Content Feedback