Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bullfrogs, Fisher Science Education™ Bio-Fresh™ Products for Science Education

Rana catesbeiana.

 View more versions of this product

Catalog No. S170750


Add to cart

Description

Description

  • Provides students with a good specimen for an in-depth dissection study of amphibians
  • Animals injected with colored latex to identify arteries and veins are available

Order Info

INJECTION KEY
Plain: Circulatory system not injected with colors.
Single: Arteries injected with red latex.
Double: Arteries red, veins blue.
Triple: Arteries red, veins blue and hepatic portal veins yellow.

Specifications

Specifications

Education
6 to 7 in.
Pail
Bullfrog
Double
15.24 to 17.78cm
Bio-Fresh™
50 Pack
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit