Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bulwark™ FR Men's Heavyweight Excel FR™ Comfortouch™ Flame Resistant Deluxe Insulated Bib Overall
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Style:
Long
Regular
Description
- Primary Closure: Zipper
- Wash Care: Industrial Laundry - Heavy Soil, Industrial Laundry - Light Soil, Home Wash
- Fabric: Shell: Flame resistant, 11 oz. (390 g/m2) Duck, Water repellent
- Blend: Shell: 88% Cotton/12% Nylon, Lining: 100% Cotton, Insulation: 100% Modacrylic
- Closure: Elasticized shoulder straps with non-conductive adjustable hardware
- Pocket: One large front bib pocket with pencil stall and two set-in front swing pockets, One watch pocket and two large hip pockets, Two-piece rule pocket and one tool pocket
- Features
- Reinforced double knees
- One large front bib pocket with pencil stall
- Two set-in front swing pockets
- Elasticized shoulder straps with non-conductive adjustable hardware
- Product Collection: Excel FR Comfortouch, Canvas
- Protection: ATPV Arc Rating: 44
- Country of Origin: Imported, Made in USA Fabric
- Fit Details: Designed to be worn over other clothing. Fit over work boots.
- Gender: Male
- Materials: Cotton/Nylon
- Brand: Bulwark™
Specifications
Closure Type | Hip Zipper with Concealed Snap |
Color | Brown |
Material | Cotton, Nylon |
Size | X-Large |
Construction | Cotton, Nylon, Modacrylic lining |
For Use With (Application) | Excellent barrier to dry particles, microorganisms and non-hazardous liquids |
Length (English) Inseam | 31.5 in. |
Length (Metric) Inseam | 80 cm |
No. of Pockets | 8 |
Size (English) Chest | 44.5 to 46.5 in. |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction