Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bulwark™ FR Men's Midweight Flame Resistant Pocketless Concealed-Gripper Work Shirt
Click to view available options
Size:
2X-Large
2X-Large (Long)
3X-Large
Large
Large (Long)
Medium
Medium (Long)
Small
X-Large
X-Large (Long)
Description
- Certifications Level 1: ASTM F1506, NFPA 2112, NFPA 70E
- Certifications Level 2: Category 2 Protection
- Hazard:
- Primary Closure: Snap/Gripper
- Wash Care: Industrial Laundry - Heavy Soil, Industrial Laundry - Light Soil, Home Wash
- Fabric: Flame resistant, 7 oz. (235 g/m2)
- Blend: 88% Cotton/12% Nylon
- Closure: Concealed gripper-front closure
- Collar: Lay-flat collar
- Features: One-piece lined cuffs with closures, Sleeve vents
- Product Collection: Excel FR Comfortouch
- Packaging: Hang tag
- Protection: ATPV Arc Rating: 8.6
- Country of Origin: Imported, Made in USA Fabric
- Fit Details: Regular Fit
- Gender: Male
- Materials: Cotton/Nylon
- Brand: Bulwark™
Specifications
Size | Large (Long) |
Gender | Men's |
Material | Cotton, Nylon |
Color | Light Blue |
Neck Style | Collar |
Closure Type | Hemmed front w/concealed gripper closures |
Resistance | Flame |
Certifications/Compliance | ASTM F1506, NFPA 2112, NFPA 70E, Category 2 Protection |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction