Bulwark™ Concealed Snap Front Cotton Lab Coats

Flame-resistant coats ideal for manufacturing industry or in university labs

Manufacturer: Red Kap Industries KLL6NV RG XXL

 View more versions of this product

Catalog No. 19-296-585

  Check Availability
Add to Cart

    Specifications

    Material Cotton - 88%, Nylon - 12%
    No. of Pockets 3.0
    Size 2X-Large
    Finish Ammonia cure and heat cure
    Gender Male
    Collar Type Lapel
    Size (English) Chest 52 to 54 in.
    Certifications/Compliance NFPA 2112
    Style Regular
    Closure Type Snap Front
    Color Navy Blue
    Construction Flame resistant

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit