Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bulwark™ Concealed Snap Front Nomex Lab Coats

Comprised of 93% Nomex Aramid, 5% Kevlar™ Aramid and 2% other fibers

Manufacturer:  Red Kap Industries KNL6RB RG S

 View more versions of this product

Catalog No. 19-296-586


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Lapel
Snap
Kevlar™ Aramid™,Nomex™ Aramid™
Small
36 to 38 in.
NFPA 2112
Royal Blue
3
Male
Regular
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit