Learn More
Bulwark™ FR Excel FR™ Comfortouch™ 7 oz. Dress Uniform Shirts
Click to view available options
Color:
Light Blue
Navy
Size:
2X-Large (Long)
2X-Large (Regular)
3X-Large (Regular)
Large (Long)
Large (Regular)
Medium (Long)
Medium (Regular)
Small (Regular)
X-Large (Long)
X-Large (Regular)
Description
- Banded, topstitched, button-down collar
- Two chest pockets with button flap closures and sewn-in pencil stall
- Placket front with button closure
- Topstitched cuff with button closure
- Tailored sleeve plackets
- Home wash and industrial wash
- Protection: Arc Rating ATPV 8.6 cal./cm2
- Available in navy and light blue colors
Specifications
Size | Large (Regular) |
Color | Navy |
Hazard Risk Category | Flame-resistant |