Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Red Kap™ Excel FR™ Flame-Resistant Contractor Coveralls

Catalog Number 19295132
Click to view available options
Size (English):
42 in.
44 in.
46 in.
48 in.
50 in.
52 in.
54 in.
56 in.
58 in.
60 in.
Height (Metric):
1.73 to 1.55 m
Size (Metric) Waist:
107.9 cm
113 cm
118.1 cm
123.1 cm
128.2 cm
133.3 cm
138.4 cm
143.5 cm
148.5 cm
153.6 cm
This item is not returnable. View return policy
This item is not returnable. View return policy

Unbeatable protection for foundries, flame-cutting and welding

  • Flame-resistant 100% cotton twill
  • Two-way concealed Nomex™ taped brass breakaway zipper; concealed snap at top of zipper and at neck
  • One-piece, top-stitched, lay-flat collar
  • Hemmed sleeves
  • Two set-in front pockets, one chest pocket, two patch hip pockets
  • Side vent openings
  • Hazard Rating Class 2
  • Arc Rating: ATPV 11.2 calories/cm2
  • Khaki or Navy Blue

Gender Men's
Material Cotton
Color Navy
Neck Style Collared
Closure Type 2-way Zipper
Resistance Flame
Collar Type Lay flat
Arc Flash Rating 10.6 Calories/cm2
Certifications/Compliance ASTM F2302, NFPA 70E™, ASTM F1506, NFPA 2112
No. of Pockets 5
Size (English) 54 in.
Size (English) Chest 52.5 to 54 in.
Size (English) Waist 54.5 in.
Size (Metric) Waist 138.4 cm
Height (English) 5.7 to 5.11 ft.
Height (Metric) 1.73 to 1.55 m
Style Regular
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit