Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bulwark™ FR Men's Midweight Excel FR™ Flame Resistant Khaki Work Pant, Waist Sizes 28 to 40 in.
Click to view available options
Gender:
Men's
Material:
Cotton
Color:
Khaki
No. of Pockets:
4
Closure Type:
Button
Product Type:
Flame-resistant Work Pants
Resistant To:
Flame
Hazard Risk Category:
HRC2
Arc Flash Rating:
ATPV 10.6 Calories/cm2
Certifications/Compliance:
Meets NFPA 2112
Description
- Primary Closure: Button
- Wash Care: Industrial Laundry - Heavy Soil, Industrial Laundry - Light Soil, Home Wash
- Fabric: Excel FR™ Flame resistant, 9 oz. (305 g/m2) Twill
- Blend: 100% Cotton
- Closure: Waistband with button closure
- Pocket: Two slack-style front pockets and left hip pocket with button closure
- Features:
- Waistband with button closure
- Two slack-style pockets
- Left hip pocket with button closure
- Product Collection: Excel FR
- Protection: ATPV Arc Rating: 11
- Country of Origin: Imported, Made in USA Fabric
- Fit Details: Sits at the waist. Regular in the seat and thigh. Slightly tapered leg.
- Gender: Male
- Materials: Cotton
- Brand: Bulwark™
Specifications
Size | 28 |
Product Type | Flame-resistant Work Pants |
Gender | Men's |
Material | Cotton |
Color | Khaki |
No. of Pockets | 4 |
Closure Type | Button |
Resistant To | Flame |
Hazard Risk Category | HRC2 |
Arc Flash Rating | ATPV 10.6 Calories/cm2 |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction