Learn More
VF Workwear Bulwark EXCEL FR™ Long Sleeve Tagless Henley Mens Shirts
Designed with flame-resistant fabric, 210g/m2 (6.25 oz.) Interlock FR 100% Cotton
Manufacturer:  VF Workwear SEL2NV RG XL
 View more versions of this product
Catalog No. 18-120-310
Add to cart
Description
Description
  • Three button placket
  • Knit henley collar
  • Rib-knit cuff
  • Single spade chest pocket
  • Tagless brand label and warning label
  • Home wash
  • Protection: Arc Rating ATPV 9.6 cal./cm2
Specifications
Specifications
Arc Rating ATPV 9.6 cal./cm2
Knit henley collar
Navy
Regular, X-Large