Bulwark™ EXCEL FR™ Long Sleeve Tagless Henley Mens Shirts
Designed with flame-resistant fabric, 210g/m2 (6.25 oz.) Interlock FR 100% Cotton
Manufacturer: Red Kap Industries SEL2NV RG 3XL
 View more versions of this product
Catalog No. 18-120-314
  Check Availability
Add to Cart
    Specifications
    Arc Flash Rating Arc Rating ATPV 9.6 cal./cm2
    Size Regular, 3X-large
    Collar Type Knit henley collar
    Color Navy
  • Description & Specifications
Related Products
Product Content Feedback