Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bulwark™ FR Men's Classic Heavyweight Excel FR™ Flame Resistant Jean, Blue Denim
Click to view available options
Gender:
Men's
Material:
Cotton
Color:
Blue
No. of Pockets:
5
Closure Type:
Button
Product Type:
Flame-resistant Jeans
Hazard Risk Category:
HRC2
Arc Flash Rating:
ATPV 20.7 Calories/cm2
Certifications/Compliance:
Meets NFPA 2112
Description
- Primary Closure: Button
- Wash Care: Industrial Laundry - Heavy Soil, Industrial Laundry - Light Soil, Home Wash
- Fabric: EXCEL FR™ Flame resistant, 14.75 oz. (500 g/m2)
- Blend: 100% Pre-Washed Cotton Denim
- Closure: Button closure
- Pocket: Five jean-style pockets
- Features:
- Sits at natural waist
- Relaxed seat and thigh
- Soft pre-wash for comfort and look
- Two-needle felled seam construction
- Product Collection: Excel FR, Denim
- Protection: ATPV Arc Rating: 21
- Country of Origin: Imported, Made in USA Fabric
- Fit Details: Sits at waist. Regular in seat and thigh. Slightly tapered leg.
- Gender: Male
- Materials: Cotton
- Brand: Bulwark™
Specifications
Size | 46 x 34 in. |
Product Type | Flame-resistant Jeans |
Gender | Men's |
Material | Cotton |
Color | Blue |
No. of Pockets | 5 |
Closure Type | Button |
Hazard Risk Category | HRC2 |
Arc Flash Rating | ATPV 20.7 Calories/cm2 |
Certifications/Compliance | Meets NFPA 2112 |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction