Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bulwark™ FR Men's Relaxed Excel FR™ Flame Resistant Jean, Dark Denim
Click to view available options
Gender:
Men's
Material:
Cotton
Color:
Blue
No. of Pockets:
5
Closure Type:
Button
Finish:
Ammonia Cure
Product Type:
Flame-resistant Jeans
Resistant To:
Flame
Hazard Risk Category:
HRC2
Arc Flash Rating:
ATPV 18.0 Calories/cm2
Certifications/Compliance:
Meets NFPA, 70E Compliant
Size (English) Inseam:
37 in.
Style:
Unhemmed
Description
- Primary Closure: Zipper
- Wash Care: Industrial Laundry - Heavy Soil, Industrial Laundry - Light Soil, Home Wash
- Fabric: EXCEL FR™ Flame resistant, 12.5 oz. (425 g/m2)
- Blend: 100% Cotton Denim
- Closure: One-piece waistband with button closure
- Pocket: Five jean-style pockets
- Features:
- Sits at natural waist
- Relaxed seat and thigh with straight leg opening
- One-piece waistband with button closure
- Product Collection: Excel FR, Denim
- Protection: ATPV Arc Rating: 18
- Country of Origin: Imported, Made in USA Fabric
- Fit Details: Sits just below the waist. Relaxed in the seat and thigh. Straight leg.
- Gender: Male
- Materials: Cotton
- Brand: Bulwark™
Specifications
Size | 50 |
Product Type | Flame-resistant Jeans |
Gender | Men's |
Material | Cotton |
Color | Blue |
No. of Pockets | 5 |
Closure Type | Button |
Finish | Ammonia Cure |
Resistant To | Flame |
Hazard Risk Category | HRC2 |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction