Bulwark™ Nomex™ IIIA HRC1 Lab Coats

Constructed with flame-resistant Nomex IIIA fabric

Manufacturer: Red Kap Industries KNL2RBRG3XL

 View more versions of this product

Catalog No. 19-295-002

  Check Availability
Add to Cart

    Specifications

    Color Royal blue
    Material Nomex™ IIIA (Nomex™ Aramid™, Kevlar™ Aramid™, other Fiber)
    No. of Pockets 1.0
    Size 3X-Large
    For Use With (Application) Chemical, oil, Gasand petrochemical industries, foundries, flame cutting and welding
    Collar Type Lapel
    Hazard Rating Class 1
    Includes Side vent openings
    Arc Flash Rating 5.6 Calories/cm2
    Resistance Flame
    Weight (English) 6 oz.
    Weight (Metric) 0.17kg
    Closure Type Button

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit