Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bulwark™ FR Nomex™ IIIA HRC1 Lab Coats
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Description
- Arc Rating: ATPV 5.6 calories/cm2
- Button front closure
- Lapel collar
- Left chest pocket
- Hemmed sleeve ends
- Side vent openings
- Royal blue
- Hazard rating class 1
Specifications
Size | 4X-Large |
Material | Nomex IIIA, Dupont™ Kevlar™ aramid fibers |
Color | Royal Blue |
Closure Type | Buttons |
Resistance | Flame |
No. of Pockets | 1 |
Collar Type | Lapel |
Hazard Rating | Class 1 |
Arc Flash Rating | 5.6 Calories/cm2 |
For Use With (Application) | Chemical, oil, Gasand petrochemical industries, foundries, flame cutting and welding |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction