Bulwark™ Nomex™ IIIA HRC1 Lab Coats

Constructed with flame-resistant Nomex IIIA fabric

Manufacturer: Red Kap Industries KNL2RBRGXXL

 View more versions of this product

Catalog No. 19-166-1072

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Material Nomex™ IIIA
    No. of Pockets 1
    Size 2X-Large
    For Use With (Application) Chemical, oil, Gasand petrochemical industries, foundries, flame cutting and welding
    Collar Type Lapel
    Hazard Rating Class 1
    Includes Side vent openings
    Arc Flash Rating 5.6 Calories/cm2
    Resistance Flame
    Weight (English) 6 oz.
    Weight (Metric) 0.17kg
    Closure Type Button
    Color Royal blue

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit