Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bulwark™ FR Men's Heavyweight Excel FR™ Flame Resistant Denim Dungaree, Stonewash
Click to view available options
Gender:
Men's
Material:
Cotton
Color:
Blue
No. of Pockets:
5
Closure Type:
Concealed button
Product Type:
Flame-resistant Dungarees
Resistant To:
Flame
Hazard Risk Category:
HRC2
Arc Flash Rating:
ATPV 20.7 Calories/cm2
Description
- Primary Closure: Zipper
- Wash Care: Industrial Laundry - Heavy Soil, Industrial Laundry - Light Soil, Home Wash
- Fabric: EXCELFR™ Flame resistant, 14.75 oz. (500 g/m2)
- Blend: 100% Cotton Denim
- Closure: Zipper fly with single metal button on waistband
- Pocket: Two deep scoop pockets, Two oversized reinforced hip pockets, One rule pocket on right leg with wrap around utility pocket
- Features:
- Sits at natural waist
- Loose seat and thigh with wider leg opening
- Two deep scoop pockets and two oversized reinforced hip pockets with rule pocket on right leg and wrap around utility pocket
- Soft pre-wash for comfort and look
- Product Collection: Excel FR, Denim
- Protection: ATPV Arc Rating: 21
- Country of Origin: Imported, Made in USA Fabric
- Fit Details: Sits at the waist. Extra room in the seat and thigh. Relaxed in the leg.
- Gender: Male
- Materials: Cotton
- Brand: Bulwark™
Specifications
Size | 40 x 30 in. |
Product Type | Flame-resistant Dungarees |
Gender | Men's |
Material | Cotton |
Color | Blue |
No. of Pockets | 5 |
Closure Type | Concealed button |
Resistant To | Flame |
Hazard Risk Category | HRC2 |
Arc Flash Rating | ATPV 20.7 Calories/cm2 |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction