Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bulwark™ FR Men's Loose Flame Resistant Jean, Stonewash
Click to view available options
Gender:
Men's
Material:
Denim
Color:
Blue
No. of Pockets:
5
Closure Type:
Zipper
Product Type:
Flame-resistant Jeans
Resistant To:
Flame
Hazard Risk Category:
HRC2
Arc Flash Rating:
ATPV 20.7 Calories/cm2
Description
- Primary Closure: Zipper
- Wash Care: Industrial Laundry - Heavy Soil, Industrial Laundry - Light Soil, Home Wash
- Fabric: EXCEL FR™ Flame resistant, 14.75 oz. (500 g/m2)
- Blend: 100% Pre-Washed Cotton Denim
- Closure: One-piece waistband with metal button closure (metal is not exposed to the skin)
- Pocket: Five jean-style pockets
- Features:
- Sits below natural waist
- Loose seat and thigh with wider leg opening than our classic fit jean
- Soft pre-wash and stone-washed fabric for comfort and look
- Product Collection: Excel FR, Denim
- Packaging: Waistband ticket
- Protection: ATPV Arc Rating: 21
- Country of Origin: Imported, Made in USA Fabric
- Fit Details: Sits just below the waist. Extra room in the seat and thigh. Fuller in the leg.
- Gender: Male
- Materials: Cotton
- Brand: Bulwark™
Specifications
Size | 30 x 30 in. |
Product Type | Flame-resistant Jeans |
Gender | Men's |
Material | Denim |
Color | Blue |
No. of Pockets | 5 |
Closure Type | Zipper |
Resistant To | Flame |
Hazard Risk Category | HRC2 |
Arc Flash Rating | ATPV 20.7 Calories/cm2 |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction