Learn More
Bulwark™ Women's Flame-Resistant Denim-Blend Curvy Fit Jeans
Manufacturer:  Red Kap Industries PMJ5DD 22 30
 View more versions of this product
Catalog No. 17-150-515
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Meets NFPA 2112
Dark Denim
5.0
22
Flame
EBT 13.0 Calories/cm2
Button
70% Cotton/30% Modacrylic
Female
30 in.