Bulwark™ Women's Flame-Resistant Denim-Blend Curvy Fit Jeans
Manufacturer: Red Kap Industries PMJ5DD 22 32
 View more versions of this product
Catalog No. 17-150-516
  Check Availability
Add to Cart
    Specifications
    Certifications Meets NFPA 2112
    No. of Pockets 5.0
    Material 70% Cotton/30% Modacrylic
    Size (English) Inseam 32 in.
    Size 22
    Arc Flash Rating EBT 13.0 Calories/cm2
    Gender Female
    Resistant To Flame
    Color Dark Denim
    Closure Type Button
  • Description & Specifications
Related Products
Product Content Feedback