Learn More
VF Workwear Bulwark Women's Flame-Resistant Denim-Blend Curvy Fit Jeans
Catalog Number 17150509
Click to view available options
Size:
14
16
18
20
22
24
Size (English) Inseam:
28 in.
30 in.
32 in.
34 in.
This item is not returnable. View return policy
This item is not returnable. View return policy
Size 18
Gender Women's
Material Cotton, Modacrylic
Color Denim
No. of Pockets 5
Closure Type Button
Resistant To Flame
Hazard Risk Category HRC2
Arc Flash Rating EBT 13.0 Calories/cm2
Certifications/Compliance Meets NFPA 2112
Size (English) Inseam 34 in.
Style Unhemmed