Learn More
Bupivacaine Hydrochloride, Monohydrate, USP, 98.5-101.5%, Spectrum™
Click to view available options
Packaging:
Amber Glass Bottle
Quantity:
5 g
B1391, 73360-54-0, C18H28N2O•HCl•H2O
Spectrum™ Bupivacaine Hydrochloride, Monohydrate, USP is the most used local anesthetic in epidurals, sympathetic nerve blocks and peripheral nerve blocks. It is listed on the World Health Organizations list of essential medicines. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle