Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bupivacaine Hydrochloride, Monohydrate, USP, 98.5-101.5%, Spectrum™

Click to view available options
Quantity:
1 g
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
73360-54-0
C18H31ClN2O2
342.91
HUCIWBPMHXGLFM-UHFFFAOYNA-N
1-butyl-N-(2,6-dimethylphenyl)piperidine-2-carboxamide hydrate hydrochloride
O.Cl.CCCCN1CCCCC1C(=O)NC1=C(C)C=CC=C1C
73360-54-0
C18H31ClN2O2
342.91
HUCIWBPMHXGLFM-UHFFFAOYNA-N
1-butyl-N-(2,6-dimethylphenyl)piperidine-2-carboxamide hydrate hydrochloride
O.Cl.CCCCN1CCCCC1C(=O)NC1=C(C)C=CC=C1C

B1391, 73360-54-0, C18H28N2O•HCl•H2O

Spectrum™ Bupivacaine Hydrochloride, Monohydrate, USP is the most used local anesthetic in epidurals, sympathetic nerve blocks and peripheral nerve blocks. It is listed on the World Health Organizations list of essential medicines. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Quantity 1 kg
Ignition Residue 0.1%
Packaging Amber Glass Bottle
CAS Max % 100%
UN Number UN1544
Percent Purity 98.5-101.5%